accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

ClCH3OOOOH3C(S)NHH3COOOCH3

CAS RN 912-60-7

IUPAC Name: (5S)-5-(4,5-dimethoxy-3-oxo-1,3-dihydro-2-benzofuran-1-yl)-4-methoxy-6-methyl-2H,5H,6H,7H,8H-[1,3]dioxolo[4,5-g]isoquinolin-6-ium chloride
Synonyms: (5S)-5-(4,5-dimethoxy-3-oxo-1H-2-benzofuran-1-yl)-4-methoxy-6-methyl-2H,5H,6H,7H,8H-[1,3]dioxolo[4,5-g]isoquinolin-6-ium chloride
Molecular Weight (g/mol): 449.88
Molecular Formula: C22H24ClNO7
InChi Key: MFLVZFXCSKVCSH-OQICONIUSA-N
SMILES: [Cl-].COC1=CC=C2C(OC(=O)C2=C1OC)[C@H]1[NH+](C)CCC2=CC3=C(OCO3)C(OC)=C12
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results