CAS RN 91223-44-8
IUPAC Name:
4-ethylphenyl 4-(4-pentylcyclohexyl)benzoate
Synonyms:
4-ethylphenyl 4-(4-pentylcyclohexyl)benzoate
Molecular Weight (g/mol):
378.56
Molecular Formula:
C26H34O2
InChi Key:
WLPXTIALVPOMAH-UHFFFAOYSA-N
SMILES:
CCCCCC1CCC(CC1)C1=CC=C(C=C1)C(=O)OC1=CC=C(CC)C=C1
1
–
1
of
1
results
1
4-Ethylphenyl trans-4-(4-n-pentylcyclohexyl)benzoate, 99%, Thermo Scientific™
CAS: 91223-44-8 Molecular Formula: C26H34O2 Molecular Weight (g/mol): 378.556 MDL Number: MFCD16879074 InChI Key: WLPXTIALVPOMAH-UHFFFAOYSA-N Synonym: 4-ethylphenyl 4-trans-4-pentylcyclohexyl benzoate,4-ethylphenyl 4-4-pentylcyclohexyl benzoate,4-ethylphenyl trans-4-4-n-pentylcyclohexyl benzoate,4-ethylphenyl 4-1s,4r-4-pentylcyclohexyl benzoate PubChem CID: 23334599 IUPAC Name: (4-ethylphenyl) 4-(4-pentylcyclohexyl)benzoate SMILES: CCCCCC1CCC(CC1)C2=CC=C(C=C2)C(=O)OC3=CC=C(C=C3)CC