accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

BOOH3CCH3H3CH3CO

CAS RN 912569-68-7

IUPAC Name: 4,4,5,5-tetramethyl-2-[3-(phenoxymethyl)phenyl]-1,3,2-dioxaborolane
Synonyms: 4,4,5,5-tetramethyl-2-[3-(phenoxymethyl)phenyl]-1,3,2-dioxaborolane
Molecular Weight (g/mol): 310.2
Molecular Formula: C19H23BO3
InChi Key: AFDZGEOQKHIWKF-UHFFFAOYSA-N
SMILES: CC1(C)OB(OC1(C)C)C1=CC(COC2=CC=CC=C2)=CC=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results