accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 912569-69-8

IUPAC Name: 4-(chloromethyl)-1-{thieno[3,2-d]pyrimidin-4-yl}piperidine
Synonyms: 4-(chloromethyl)-1-{thieno[3,2-d]pyrimidin-4-yl}piperidine
Molecular Weight (g/mol): 267.78
Molecular Formula: C12H14ClN3S
InChi Key: FWDQRROENFHLAI-UHFFFAOYSA-N
SMILES: ClCC1CCN(CC1)C1=C2SC=CC2=NC=N1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results