accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

FFFH2NOHOHCl

CAS RN 91291-66-6

IUPAC Name: 3-amino-4,4,4-trifluorobutanoic acid hydrochloride
Synonyms: 3-amino-4,4,4-trifluorobutanoic acid hydrochloride
Molecular Weight (g/mol): 193.55
Molecular Formula: C4H7ClF3NO2
InChi Key: MRMARVCQZOYQPI-UHFFFAOYNA-N
SMILES: Cl.NC(CC(O)=O)C(F)(F)F

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (1)