accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

H3COHO

CAS RN 91328-77-7

IUPAC Name: 4-ethylcyclohexane-1-carboxylic acid
Synonyms: 4-ethylcyclohexane-1-carboxylic acid
Molecular Weight (g/mol): 156.23
Molecular Formula: C9H16O2
InChi Key: UNROFSAOTBVBBT-UHFFFAOYSA-N
SMILES: CCC1CCC(CC1)C(O)=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results