accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 913835-28-6

BSOONHHOOH

CAS RN 913835-28-6

IUPAC Name: [3-(cyclopropylsulfamoyl)phenyl]boronic acid
Synonyms: 3-(cyclopropylsulfamoyl)phenylboronic acid
Molecular Weight (g/mol): 241.07
Molecular Formula: C9H12BNO4S
InChi Key: WIZSPBQIRRAOMO-UHFFFAOYSA-N
SMILES: OB(O)C1=CC(=CC=C1)S(=O)(=O)NC1CC1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results