accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 913835-58-2

BOHNOH3CFHOOH

CAS RN 913835-58-2

IUPAC Name: [3-fluoro-4-(methoxycarbamoyl)phenyl]boronic acid
Synonyms: 3-fluoro-4-(methoxycarbamoyl)phenylboronic acid
Molecular Weight (g/mol): 212.97
Molecular Formula: C8H9BFNO4
InChi Key: RZYYOLUTTNFCAU-UHFFFAOYSA-N
SMILES: CONC(=O)C1=C(F)C=C(C=C1)B(O)O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results