accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 913835-81-1

BNOOH3CH3CCH3OCH3HOHO

CAS RN 913835-81-1

IUPAC Name: {1-[(tert-butoxy)carbonyl]-7-methoxy-1H-indol-2-yl}boronic acid
Synonyms: 1-(tert-butoxycarbonyl)-7-methoxyindol-2-ylboronic acid
Molecular Weight (g/mol): 291.11
Molecular Formula: C14H18BNO5
InChi Key: JYRDIGLHLIDRNE-UHFFFAOYSA-N
SMILES: COC1=C2N(C(=O)OC(C)(C)C)C(=CC2=CC=C1)B(O)O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results