accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 913836-14-3

BClCH3HOOH

CAS RN 913836-14-3

IUPAC Name: (3-chloro-5-methylphenyl)boronic acid
Synonyms: 3-chloro-5-methylphenylboronic acid
Molecular Weight (g/mol): 170.4
Molecular Formula: C7H8BClO2
InChi Key: QXHLBINWDJITMJ-UHFFFAOYSA-N
SMILES: CC1=CC(Cl)=CC(=C1)B(O)O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results