accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 913836-27-8

IUPAC Name: 2-[4-(2-bromoethoxy)phenyl]-4,4,5,5-tetramethyl-1,3,2-dioxaborolane
Synonyms: 2-[4-(2-bromoethoxy)phenyl]-4,4,5,5-tetramethyl-1,3,2-dioxaborolane
Molecular Weight (g/mol): 327.03
Molecular Formula: C14H20BBrO3
InChi Key: NTTOMYNPPZBRBG-UHFFFAOYSA-N
SMILES: CC1(C)OB(OC1(C)C)C1=CC=C(OCCBr)C=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results