accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

NNOH2NFFF

CAS RN 914348-10-0

IUPAC Name: 2-(trifluoromethyl)pyrimidine-4-carboxamide
Synonyms: 2-(trifluoromethyl)pyrimidine-4-carboxamide
Molecular Weight (g/mol): 191.11
Molecular Formula: C6H4F3N3O
InChi Key: LBNILUATVOCQRF-UHFFFAOYSA-N
SMILES: NC(=O)C1=NC(=NC=C1)C(F)(F)F

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results