accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 916204-05-2

IUPAC Name: 4-[4-({4'-chloro-[1,1'-biphenyl]-2-yl}methyl)piperazin-1-yl]benzoic acid
Synonyms: 4-[4-({4'-chloro-[1,1'-biphenyl]-2-yl}methyl)piperazin-1-yl]benzoic acid
Molecular Weight (g/mol): 406.91
Molecular Formula: C24H23ClN2O2
InChi Key: FSHGEKXJPYDBSO-UHFFFAOYSA-N
SMILES: OC(=O)C1=CC=C(C=C1)N1CCN(CC2=CC=CC=C2C2=CC=C(Cl)C=C2)CC1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results