accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

ClONH2FFF

CAS RN 916420-45-6

IUPAC Name: 2-chloro-6-(trifluoromethyl)benzamide
Synonyms: 2-chloro-6-(trifluoromethyl)benzamide
Molecular Weight (g/mol): 223.58
Molecular Formula: C8H5ClF3NO
InChi Key: JUAICIUGQMFFPO-UHFFFAOYSA-N
SMILES: NC(=O)C1=C(C=CC=C1Cl)C(F)(F)F

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results