accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

FFFOClFFF

CAS RN 916420-48-9

IUPAC Name: 2-chloro-4,6-bis(trifluoromethyl)benzaldehyde
Synonyms: 2-chloro-4,6-bis(trifluoromethyl)benzaldehyde
Molecular Weight (g/mol): 276.56
Molecular Formula: C9H3ClF6O
InChi Key: FFSCXVIOXOGOIT-UHFFFAOYSA-N
SMILES: FC(F)(F)C1=CC(=C(C=O)C(Cl)=C1)C(F)(F)F

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results