CAS RN 916420-50-3
IUPAC Name:
1-(bromomethyl)-3-fluoro-2-methoxybenzene
Synonyms:
1-(bromomethyl)-3-fluoro-2-methoxybenzene
Molecular Weight (g/mol):
219.05
Molecular Formula:
C8H8BrFO
InChi Key:
OYXRYBDZSXMMPU-UHFFFAOYSA-N
SMILES:
COC1=C(CBr)C=CC=C1F
1
–
1
of
1
results
1
3-Fluoro-2-methoxybenzyl bromide, 97%, Thermo Scientific™
CAS: 916420-50-3 Molecular Formula: C8H8BrFO Molecular Weight (g/mol): 219.053 MDL Number: MFCD09025370 InChI Key: OYXRYBDZSXMMPU-UHFFFAOYSA-N Synonym: 3-fluoro-2-methoxybenzyl bromide,1-bromomethyl-3-fluoro-2-methoxybenzene PubChem CID: 46737536 IUPAC Name: 1-(bromomethyl)-3-fluoro-2-methoxybenzene SMILES: COC1=C(C=CC=C1F)CBr