accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 916420-52-5

CH3OClOFFF

CAS RN 916420-52-5

IUPAC Name: 3-methyl-5-(trifluoromethoxy)benzoyl chloride
Synonyms: 3-methyl-5-(trifluoromethoxy)benzoyl chloride
Molecular Weight (g/mol): 238.59
Molecular Formula: C9H6ClF3O2
InChi Key: CCKDSMKGHHIBFA-UHFFFAOYSA-N
SMILES: CC1=CC(OC(F)(F)F)=CC(=C1)C(Cl)=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results