accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

H3CNCH3OH2N

CAS RN 916766-87-5

IUPAC Name: 1-{2-[3-(dimethylamino)propoxy]phenyl}methanamine
Synonyms: 1-{2-[3-(dimethylamino)propoxy]phenyl}methanamine
Molecular Weight (g/mol): 208.31
Molecular Formula: C12H20N2O
InChi Key: CWYPVGNYPWEJLK-UHFFFAOYSA-N
SMILES: CN(C)CCCOC1=CC=CC=C1CN

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results