accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 916766-91-1

IUPAC Name: 1-{thieno[3,2-d]pyrimidin-4-yl}piperidine-4-carbaldehyde
Synonyms: 1-{thieno[3,2-d]pyrimidin-4-yl}piperidine-4-carbaldehyde
Molecular Weight (g/mol): 247.32
Molecular Formula: C12H13N3OS
InChi Key: YGMBYHWGCCTCNN-UHFFFAOYSA-N
SMILES: O=CC1CCN(CC1)C1=C2SC=CC2=NC=N1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results