accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 916766-99-9

IUPAC Name: 3-hydroxy-5-{[(prop-2-en-1-yloxy)carbonyl]amino}benzoic acid
Synonyms: 3-hydroxy-5-{[(prop-2-en-1-yloxy)carbonyl]amino}benzoic acid
Molecular Weight (g/mol): 237.21
Molecular Formula: C11H11NO5
InChi Key: MYOCYAGTJQROGX-UHFFFAOYSA-N
SMILES: OC(=O)C1=CC(NC(=O)OCC=C)=CC(O)=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results