accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 916791-97-4

CH3OONNHH3CH3CCH3

CAS RN 916791-97-4

IUPAC Name: ethyl 5-tert-butyl-1H-pyrazole-3-carboxylate
Synonyms: ethyl 5-tert-butyl-1H-pyrazole-3-carboxylate
Molecular Weight (g/mol): 196.25
Molecular Formula: C10H16N2O2
InChi Key: RCXMILPYEKVQLB-UHFFFAOYSA-N
SMILES: CCOC(=O)C1=NNC(=C1)C(C)(C)C

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results