accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

H3CLi

CAS RN 917-54-4

IUPAC Name: lithium(1+) methanide
Synonyms: methyllithium
Molecular Weight (g/mol): 21.98
Molecular Formula: CH3Li
InChi Key: IHLVCKWPAMTVTG-UHFFFAOYSA-N
SMILES: [Li+].[CH3-]
Quantity 
  • (3)
  • (3)
Percent Purity 
  • (2)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results