accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 91742-21-1

IUPAC Name: 1,1,1-trifluoro-N-trifluoromethanesulfonylmethanesulfonamide sodium
Synonyms: 1,1,1-trifluoro-N-trifluoromethanesulfonylmethanesulfonamide sodium
Molecular Weight (g/mol): 304.13
Molecular Formula: C2HF6NNaO4S2
InChi Key: QXZNUMVOKMLCEX-UHFFFAOYSA-N
SMILES: [Na].FC(F)(F)S(=O)(=O)NS(=O)(=O)C(F)(F)F
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results