accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CH3OSiNH2OH3COH3C

CAS RN 919-30-2

IUPAC Name: (3-aminopropyl)triethoxysilane
Synonyms: aminopropyl triethoxysilane
Molecular Weight (g/mol): 221.37
Molecular Formula: C9H23NO3Si
InChi Key: WYTZZXDRDKSJID-UHFFFAOYSA-N
SMILES: CCO[Si](CCCN)(OCC)OCC
Quantity 
  • (3)
  • (1)
  • (1)
  • (1)
  • (2)
  • (1)
  • (2)
  • (1)
Percent Purity 
  • (3)
  • (6)
Boiling Point 
  • (6)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (3)
  • (1)
  • (1)
  • (1)
  • (2)
  • (1)
  • (2)
  • (1)
  • (3)
  • (6)
  • (6)
  • (6)