accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

H3CCH3HOO

CAS RN 91968-71-7

IUPAC Name: 2-(benzyloxy)-2-methylpropan-1-ol
Synonyms: 2-(benzyloxy)-2-methylpropan-1-ol
Molecular Weight (g/mol): 180.25
Molecular Formula: C11H16O2
InChi Key: FRHJVAJQSPVSKV-UHFFFAOYSA-N
SMILES: CC(C)(CO)OCC1=CC=CC=C1
Boiling Point 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results