accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

ONOOOO

CAS RN 91990-88-4

IUPAC Name: 2,5-dioxopyrrolidin-1-yl 4-benzoylbenzoate
Synonyms: 2,5-dioxopyrrolidin-1-yl 4-benzoylbenzoate
Molecular Weight (g/mol): 323.3
Molecular Formula: C18H13NO5
InChi Key: MVQNJLJLEGZFGP-UHFFFAOYSA-N
SMILES: O=C(ON1C(=O)CCC1=O)C1=CC=C(C=C1)C(=O)C1=CC=CC=C1
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results