accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

FFFSNH

CAS RN 92-30-8

IUPAC Name: 2-(trifluoromethyl)-10H-phenothiazine
Synonyms: trifluoromethylphenothiazine
Molecular Weight (g/mol): 267.27
Molecular Formula: C13H8F3NS
InChi Key: RKGYJVASTMCSHZ-UHFFFAOYSA-N
SMILES: FC(F)(F)C1=CC=C2SC3=CC=CC=C3NC2=C1
Molecular Weight (g/mol) 
  • (2)
  • (2)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results