accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 92-32-0

ClH3CNCH3ONCH3CH3

CAS RN 92-32-0

IUPAC Name: 3,6-bis(dimethylamino)-10λ⁴-xanthen-10-ylium chloride
Synonyms: 3,6-bis(dimethylamino)-10λ⁴-xanthen-10-ylium chloride
Molecular Weight (g/mol): 302.8
Molecular Formula: C17H19ClN2O
InChi Key: INCIMLINXXICKS-UHFFFAOYSA-M
SMILES: [Cl-].CN(C)C1=CC2=[O+]C3=CC(=CC=C3C=C2C=C1)N(C)C
Quantity 
  • (3)
  • (1)
  • (1)
  • (1)
  • (1)
  • (4)
Color 
  • (8)
  • (2)
Grade 
  • (5)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (5)
  • (5)
  • (1)
  • (3)
  • (1)
  • (1)
  • (1)
  • (1)
  • (4)
  • (8)
  • (2)
  • (5)