accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 92-61-5

IUPAC Name: 7-hydroxy-6-methoxy-2H-chromen-2-one
Synonyms: scopoletin 东莨菪亭 acid, chrysotropic 6-methylesculetin 6-O-methylesculetin scopoletol gelseminic acid β-methylesculetin chrysotropic acid acid, gelseminic
Molecular Weight (g/mol): 192.17
Molecular Formula: C10H8O4
InChi Key: RODXRVNMMDRFIK-UHFFFAOYSA-N
SMILES: COC1=C(O)C=C2OC(=O)C=CC2=C1
Percent Purity 
  • (3)
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
Physical Form 
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (3)
  • (1)
  • (3)
  • (3)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (3)