accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

BrBr

CAS RN 92-86-4

IUPAC Name: 4,4'-dibromo-1,1'-biphenyl
Synonyms: 4,4'-dibromobiphenyl
Molecular Weight (g/mol): 312
Molecular Formula: C12H8Br2
InChi Key: HQJQYILBCQPYBI-UHFFFAOYSA-N
SMILES: BrC1=CC=C(C=C1)C1=CC=C(Br)C=C1
Solvent or Matrix 
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
Boiling Point 
  • (5)
Concentration 
  • (2)
  • (3)
Percent Purity 
  • (2)
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (5)
  • (1)
  • (3)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (5)
  • (2)
  • (3)
  • (2)
  • (3)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (3)