accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

OHOH

CAS RN 92-88-6

IUPAC Name: [1,1'-biphenyl]-4,4'-diol
Synonyms: 4,4'-dihydroxybiphenyl 4,4'-dioxydiphenol 4,4'-dioxydiphenyl dihydroxybiphenyl p,p'-diphenol 4,4'-dihydroxydiphenyl asm dod DOD antioxidant dod 44'BP cpd
Molecular Weight (g/mol): 186.21
Molecular Formula: C12H10O2
InChi Key: VCCBEIPGXKNHFW-UHFFFAOYSA-N
SMILES: OC1=CC=C(C=C1)C1=CC=C(O)C=C1
Percent Purity 
  • (1)
  • (3)
  • (3)
Quantity 
  • (1)
  • (2)
  • (2)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results