accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 92-92-2

HOO

CAS RN 92-92-2

IUPAC Name: [1,1'-biphenyl]-4-carboxylic acid
Synonyms: [1,1'-biphenyl]-4-carboxylic acid
Molecular Weight (g/mol): 198.22
Molecular Formula: C13H10O2
InChi Key: NNJMFJSKMRYHSR-UHFFFAOYSA-N
SMILES: OC(=O)C1=CC=C(C=C1)C1=CC=CC=C1
Molecular Weight (g/mol) 
  • (6)
  • (3)
Quantity 
  • (2)
  • (2)
  • (1)
  • (1)
  • (2)
  • (1)
Physical Form 
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (3)
  • (3)
  • (6)
  • (3)
  • (2)
  • (2)
  • (1)
  • (1)
  • (2)
  • (1)
  • (3)