accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

LiH3CH3CCH2

CAS RN 920-36-5

IUPAC Name: lithium(1+) 2-methylpropan-1-ide
Synonyms: lithium(1+) 2-methylpropan-1-ide
Molecular Weight (g/mol): 64.06
Molecular Formula: C4H9Li
InChi Key: CCZVEWRRAVASGL-UHFFFAOYSA-N
SMILES: [Li+].CC(C)[CH2-]
Percent Purity 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (1)
  • (1)