accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CH3H3CNOHO(?)

CAS RN 92028-57-4

IUPAC Name: 2-(2,5-dimethyl-1H-pyrrol-1-yl)benzoic acid
Synonyms: 2-(2,5-dimethylpyrrol-1-yl)benzoic acid
Molecular Weight (g/mol): 215.25
Molecular Formula: C13H13NO2
InChi Key: ZLYUUANOICYAAL-UHFFFAOYSA-N
SMILES: CC1=CC=C(C)N1C1=CC=CC=C1C(O)=O
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results