accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

H3COOOOOH3COSOOFFFOOCH3OOCH3

CAS RN 92051-23-5

IUPAC Name: [3,4,6-tris(acetyloxy)-5-(trifluoromethanesulfonyloxy)oxan-2-yl]methyl acetate
Synonyms: [3,4,6-tris(acetyloxy)-5-(trifluoromethanesulfonyloxy)oxan-2-yl]methyl acetate
Molecular Weight (g/mol): 480.36
Molecular Formula: C15H19F3O12S
InChi Key: OIBDVHSTOUGZTJ-UHFFFAOYNA-N
SMILES: CC(=O)OCC1OC(OC(C)=O)C(OS(=O)(=O)C(F)(F)F)C(OC(C)=O)C1OC(C)=O
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results