accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 921-01-7

IUPAC Name: (2S)-2-amino-3-sulfanylpropanoic acid
Synonyms: D-cysteine ecolan cysteinum D-cystein cisteinum L cysteine half cystine dl-cysteina ccris 912 D-zystein
Molecular Weight (g/mol): 121.15
Molecular Formula: C3H7NO2S
InChi Key: XUJNEKJLAYXESH-UWTATZPHSA-N
SMILES: N[C@H](CS)C(O)=O
Molecular Weight (g/mol) 
  • (3)
  • (3)
Quantity 
  • (3)
  • (1)
  • (2)
Percent Purity 
  • (1)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (3)
  • (3)
  • (3)
  • (3)
  • (1)
  • (2)
  • (1)
  • (2)