accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 92149-07-0

IUPAC Name: 4,7-dimethoxy-1,10-phenanthroline
Synonyms: 4,7-dimethoxy-1,10-phenanthroline
Molecular Weight (g/mol): 240.26
Molecular Formula: C14H12N2O2
InChi Key: ZPGVCQYKXIQWTP-UHFFFAOYSA-N
SMILES: COC1=C2C=CC3=C(OC)C=CN=C3C2=NC=C1
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results