accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 921938-51-4

IUPAC Name: 2,3,4,5,6-pentafluorophenyl 4-(morpholin-4-yl)benzoate
Synonyms: 2,3,4,5,6-pentafluorophenyl 4-(morpholin-4-yl)benzoate
Molecular Weight (g/mol): 373.28
Molecular Formula: C17H12F5NO3
InChi Key: UJYQDNSLPIRXRT-UHFFFAOYSA-N
SMILES: FC1=C(F)C(F)=C(OC(=O)C2=CC=C(C=C2)N2CCOCC2)C(F)=C1F

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results