accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 924-49-2

IUPAC Name: (3S)-4-azaniumyl-3-hydroxybutanoate
Synonyms: 4-amino-3-hydroxybutyric acid
Molecular Weight (g/mol): 119.12
Molecular Formula: C4H9NO3
InChi Key: YQGDEPYYFWUPGO-VKHMYHEASA-N
SMILES: [NH3+]C[C@@H](O)CC([O-])=O
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results