accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 92404-63-2

IUPAC Name: 3-bromofuro[3,2-b]pyridine
Synonyms: 3-bromofuro[3,2-b]pyridine
Molecular Weight (g/mol): 198.02
Molecular Formula: C7H4BrNO
InChi Key: KHMAAFDMZSYVEF-UHFFFAOYSA-N
SMILES: BrC1=COC2=CC=CN=C12
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results