accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 92557-80-7

IUPAC Name: 2,5-dioxopyrrolidin-1-yl 3',6'-dihydroxy-3-oxo-3H-spiro[2-benzofuran-1,9'-xanthene]-5-carboxylate
Synonyms: 2,5-dioxopyrrolidin-1-yl 3',6'-dihydroxy-3-oxospiro[2-benzofuran-1,9'-xanthene]-5-carboxylate
Molecular Weight (g/mol): 473.39
Molecular Formula: C25H15NO9
InChi Key: GECIDMICWWDIBO-UHFFFAOYSA-N
SMILES: OC1=CC2=C(C=C1)C1(OC(=O)C3=C1C=CC(=C3)C(=O)ON1C(=O)CCC1=O)C1=C(O2)C=C(O)C=C1
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results