accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 926921-59-7

IUPAC Name: 2,3,4,5,6-pentafluorophenyl 6-(thiophen-2-yl)pyridine-3-carboxylate
Synonyms: 2,3,4,5,6-pentafluorophenyl 6-(thiophen-2-yl)pyridine-3-carboxylate
Molecular Weight (g/mol): 371.28
Molecular Formula: C16H6F5NO2S
InChi Key: UYAXHNWJTLZXTM-UHFFFAOYSA-N
SMILES: FC1=C(F)C(F)=C(OC(=O)C2=CN=C(C=C2)C2=CC=CS2)C(F)=C1F

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results