accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 927391-09-1

CH3SnH3CCH3SNOCH3

CAS RN 927391-09-1

IUPAC Name: 2-methoxy-4-(tributylstannyl)-1,3-thiazole
Synonyms: 2-methoxy-4-(tributylstannyl)-1,3-thiazole
Molecular Weight (g/mol): 404.2
Molecular Formula: C16H31NOSSn
InChi Key: JCZRGWAGRPABLT-UHFFFAOYSA-N
SMILES: CCCC[Sn](CCCC)(CCCC)C1=CSC(OC)=N1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results