accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 928-95-0

CH3OH(E)

CAS RN 928-95-0

IUPAC Name: (2E)-hex-2-en-1-ol
Synonyms: trans-2-hexenol
Molecular Weight (g/mol): 100.16
Molecular Formula: C6H12O
InChi Key: ZCHHRLHTBGRGOT-SNAWJCMRSA-N
SMILES: CCC\C=C\CO
Boiling Point 
  • (3)
  • (2)
Percent Purity 
  • (2)
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (3)
  • (3)
  • (2)
  • (2)
  • (3)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)