accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CH3HO(Z)

CAS RN 928-96-1

IUPAC Name: (3Z)-hex-3-en-1-ol
Synonyms: 3-hexenol 3-hexen-1-ol, (Z)-isomer leaf alcohol 3-hexen-1-ol, (Z)- cis-3-hexenol (natural) 3-hexen-1-ol, (E)- 3-hexen-1-ol, (e)-isomer (E)-3-hexen-1-ol 3-hexen-1-ol, (z)- (z)-isomer 3-hexen-1-ol
Molecular Weight (g/mol): 100.16
Molecular Formula: C6H12O
InChi Key: UFLHIIWVXFIJGU-ARJAWSKDSA-N
SMILES: CC\C=C/CCO
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
Boiling Point 
  • (3)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (3)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (3)
  • (2)
  • (2)