accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 929-98-6

IUPAC Name: 1-(nonylsulfanyl)nonane
Synonyms: 1-(nonylsulfanyl)nonane
Molecular Weight (g/mol): 286.56
Molecular Formula: C18H38S
InChi Key: KMKSVAGOBVUFRO-UHFFFAOYSA-N
SMILES: CCCCCCCCCSCCCCCCCCC
Boiling Point 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results