accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 93-07-2

IUPAC Name: 3,4-dimethoxybenzoic acid
Synonyms: veratric acid veratrumenoic acid dimethylprotocatechuic acid
Molecular Weight (g/mol): 182.18
Molecular Formula: C9H10O4
InChi Key: DAUAQNGYDSHRET-UHFFFAOYSA-N
SMILES: COC1=CC=C(C=C1OC)C(O)=O
Molecular Weight (g/mol) 
  • (3)
  • (3)
  • (3)
Percent Purity 
  • (3)
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
Physical Form 
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results