accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 93-14-1

IUPAC Name: 3-(2-methoxyphenoxy)propane-1,2-diol
Synonyms: guaifenesin guaifenesin (inn)
Molecular Weight (g/mol): 198.22
Molecular Formula: C10H14O4
InChi Key: HSRJKNPTNIJEKV-UHFFFAOYNA-N
SMILES: COC1=CC=CC=C1OCC(O)CO
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
Boiling Point 
  • (3)
Certifications/Compliance 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (3)
  • (3)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (3)
  • (3)
  • (1)
  • (1)
  • (1)