accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 93-35-6

IUPAC Name: 7-hydroxy-2H-chromen-2-one
Synonyms: umbelliferone COUMARIN7hydroxy 14c-labeled 7-hydroxycoumarin β-umbelliferone 14C-labeled 7-hydroxycoumarin hydrangin skimmetin 7-hydroxycoumarin, 14C-labeled 7-hydroxycoumarin, 14c-labeled umbelliferon
Molecular Weight (g/mol): 162.14
Molecular Formula: C9H6O3
InChi Key: ORHBXUUXSCNDEV-UHFFFAOYSA-N
SMILES: OC1=CC=C2C=CC(=O)OC2=C1
Percent Purity 
  • (3)
  • (2)
  • (2)
Quantity 
  • (1)
  • (2)
  • (1)
  • (1)
  • (2)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (3)
  • (3)
  • (2)
  • (2)
  • (1)
  • (2)
  • (1)
  • (1)
  • (2)
  • (2)