accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

H3COCH3HO

CAS RN 93-51-6

IUPAC Name: 2-methoxy-4-methylphenol
Synonyms: creosol
Molecular Weight (g/mol): 138.17
Molecular Formula: C8H10O2
InChi Key: PETRWTHZSKVLRE-UHFFFAOYSA-N
SMILES: COC1=CC(C)=CC=C1O
Molecular Weight (g/mol) 
  • (1)
  • (4)
  • (2)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
Boiling Point 
  • (3)
  • (2)
Percent Purity 
  • (3)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (3)
  • (1)
  • (4)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (3)
  • (2)
  • (3)
  • (2)
  • (2)
  • (1)
  • (1)